Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334858.1 | NP_001321744.1 | 14 - 15 | 2414 | 76 | |
NM_001334859.1 | NP_001321746.1 | 15 - 16 | 2337 | 76 | ||
NM_001334860.1 | NP_001321747.1 | 15 - 16 | 2330 | 76 | ||
NM_148667.2 | NP_683508.1 | 16 - 17 | 2170 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |