Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCG38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339062.1 | NP_001319673.1 | 1 - 2 | 304 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |