Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BZO2H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340389.1 | NP_001329302.1 | 2 - 3 | 585 | 83 | |
NM_178959.3 | NP_849290.1 | 2 - 3 | 576 | 83 | ||
GenBank mRNA | AF310222.1 | - | 2 - 3 | 523 | 83 | |
BT025558.1 | - | 2 - 3 | 499 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |