Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATKRS-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112003.4 | NP_187777.1 | 6 - 7 | 710 | 106 | |
GenBank mRNA | AF125574.1 | - | 6 - 7 | 693 | 106 | |
AY099551.1 | - | 6 - 7 | 671 | 106 | ||
BT002123.1 | - | 6 - 7 | 607 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |