Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATFP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112005.4 | NP_187779.1 | 5 - 6 | 465 | 77 | |
GenBank mRNA | AK118150.1 | - | 5 - 6 | 352 | 77 | |
BT005576.1 | - | 4 - 5 | 245 | 77 | ||
U64911.1 | - | 5 - 6 | 328 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |