Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337960.1 | NP_001327533.1 | 1 - 2 | 315 | 76 | |
NM_112023.2 | NP_187796.1 | 1 - 2 | 315 | 76 | ||
GenBank mRNA | AK227614.1 | - | 1 - 2 | 315 | 76 | |
BT008302.1 | - | 1 - 2 | 244 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |