Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nek7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337978.1 | NP_001327416.1 | 2 - 3 | 645 | 222 | |
NM_112055.5 | NP_187827.1 | 2 - 3 | 576 | 145 | ||
GenBank mRNA | BT015419.1 | - | 2 - 3 | 195 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |