Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nek7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202938.1 | NP_001189867.1 | 8 - 9 | 960 | 130 | |
NM_001337978.1 | NP_001327416.1 | 6 - 7 | 1007 | 130 | ||
NM_001337979.1 | NP_001327417.1 | 7 - 8 | 1221 | 130 | ||
NM_112055.5 | NP_187827.1 | 7 - 8 | 861 | 130 | ||
GenBank mRNA | BT015419.1 | - | 7 - 8 | 480 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |