Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337980.1 | NP_001327414.1 | 13 - 14 | 1243 | 158 | |
NM_001337981.1 | NP_001327415.1 | 13 - 14 | 1249 | 158 | ||
NM_112056.3 | NP_187828.1 | 12 - 13 | 1209 | 158 | ||
GenBank mRNA | BT012565.1 | - | 12 - 13 | 1184 | 158 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |