Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337980.1 | NP_001327414.1 | 4 - 5 | 422 | 80 | |
NM_001337981.1 | NP_001327415.1 | 4 - 5 | 428 | 80 | ||
NM_112056.3 | NP_187828.1 | 3 - 4 | 388 | 80 | ||
GenBank mRNA | BT012565.1 | - | 3 - 4 | 363 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |