Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035601.1 | NP_001030678.1 | 5 - 6 | 354 | 60 | |
NM_001084672.1 | NP_001078141.1 | 6 - 7 | 594 | 60 | ||
NM_001337984.1 | NP_001326019.1 | 5 - 6 | 851 | 60 | ||
NM_001337985.1 | NP_001326020.1 | 6 - 7 | 676 | 60 | ||
NM_112061.6 | NP_566415.3 | 5 - 6 | 584 | 60 | ||
NM_202563.3 | NP_974292.1 | 6 - 7 | 658 | 60 | ||
NM_202564.2 | NP_974293.1 | 4 - 5 | 693 | 60 | ||
GenBank mRNA | AB493610.1 | - | 4 - 5 | 279 | 60 | |
AJ320540.1 | - | 6 - 7 | 513 | 60 | ||
BT029515.1 | - | 4 - 5 | 279 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |