Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035601.1 | NP_001030678.1 | 6 - 7 | 404 | 68 | |
NM_001084672.1 | NP_001078141.1 | 7 - 8 | 644 | 68 | ||
NM_001337984.1 | NP_001326019.1 | 6 - 7 | 901 | 68 | ||
NM_001337985.1 | NP_001326020.1 | 7 - 8 | 726 | 68 | ||
NM_112061.6 | NP_566415.3 | 6 - 7 | 634 | 68 | ||
NM_202563.3 | NP_974292.1 | 7 - 8 | 708 | 68 | ||
NM_202564.2 | NP_974293.1 | 5 - 6 | 743 | 68 | ||
GenBank mRNA | AB493610.1 | - | 5 - 6 | 329 | 68 | |
AJ320540.1 | - | 7 - 8 | 563 | 68 | ||
BT029515.1 | - | 5 - 6 | 329 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |