Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202939.1 | NP_001189868.1 | 5 - 6 | 724 | 93 | |
NM_112064.5 | NP_566417.3 | 5 - 6 | 913 | 93 | ||
GenBank mRNA | AF245395.1 | - | 5 - 6 | 631 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |