Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112075.3 | NP_566423.1 | 1 | 1462 | 108 | |
GenBank mRNA | AY056283.1 | - | 1 | 1450 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |