Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035603.1 | NP_001030680.1 | 5 - 6 | 351 | 79 | |
NM_001337995.1 | NP_001319531.1 | 7 - 8 | 554 | 79 | ||
NM_001337996.1 | NP_001326331.1 | 6 - 7 | 577 | 79 | ||
NM_112088.3 | NP_187859.2 | 6 - 7 | 676 | 79 | ||
GenBank mRNA | BT010931.1 | - | 6 - 7 | 435 | 79 | |
BT020600.1 | - | 5 - 6 | 370 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |