Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IAA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119006.3 | NP_194593.1 | 4 - 5 | 877 | 101 | |
GenBank mRNA | AK118377.1 | - | 4 - 5 | 863 | 101 | |
U18413.1 | - | 4 - 5 | 767 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |