Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119015.4 | NP_194600.2 | 19 - 20 | 2426 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |