Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HXK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119057.4 | NP_194642.1 | 2 - 3 | 753 | 114 | |
GenBank mRNA | AK230265.1 | - | 2580 | 114 | ||
AY075658.1 | - | 2 - 3 | 643 | 114 | ||
AY124809.1 | - | 2 - 3 | 417 | 114 | ||
U28214.1 | - | 2 - 3 | 614 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |