Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341965.1 | NP_001328427.1 | 2 - 3 | 431 | 156 | |
NM_119078.5 | NP_194662.2 | 2 - 3 | 539 | 156 | ||
GenBank mRNA | AY063817.1 | - | 2 - 3 | 272 | 156 | |
AY096457.1 | - | 2 - 3 | 155 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |