Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341965.1 | NP_001328427.1 | 4 - 5 | 590 | 96 | |
NM_119078.5 | NP_194662.2 | 4 - 5 | 698 | 96 | ||
GenBank mRNA | AY063817.1 | - | 4 - 5 | 431 | 96 | |
AY096457.1 | - | 4 - 5 | 314 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |