Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARF16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119154.5 | NP_567841.1 | 2 - 3 | 1288 | 75 | |
GenBank mRNA | AY059792.1 | - | 2 - 3 | 1243 | 75 | |
AY091198.1 | - | 1 - 2 | 1091 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |