Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VEL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203942.1 | NP_001190871.1 | 3 - 4 | 707 | 109 | |
NM_119166.5 | NP_194749.1 | 3 - 4 | 707 | 109 | ||
NM_179140.2 | NP_849471.1 | 2 - 3 | 613 | 109 | ||
NM_202910.4 | NP_974639.1 | 3 - 4 | 707 | 109 | ||
GenBank mRNA | AB050977.1 | - | 3 - 4 | 673 | 109 | |
BT002499.1 | - | 2 - 3 | 567 | 109 | ||
BT008428.1 | - | 1 - 2 | 358 | 109 | ||
EF064792.1 | - | 3 - 4 | 674 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |