Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNGC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119182.4 | NP_194765.2 | 5 - 6 | 1671 | 83 | |
GenBank mRNA | AK227176.1 | - | 5 - 6 | 1648 | 83 | |
AY124000.1 | - | 5 - 6 | 1647 | 83 | ||
BT002284.1 | - | 5 - 6 | 1431 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |