Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TET9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119189.1 | NP_194772.1 | 1 - 2 | 563 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |