Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119227.4 | NP_567854.1 | 2 - 3 | 705 | 79 | |
GenBank mRNA | AY050958.1 | - | 2 - 3 | 400 | 79 | |
AY133774.1 | - | 2 - 3 | 377 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |