Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HHP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119232.3 | NP_194814.2 | 1 - 2 | 456 | 90 | |
NM_202914.1 | NP_974643.1 | 1 - 2 | 396 | 90 | ||
GenBank mRNA | AY267331.1 | - | 1 - 2 | 229 | 90 | |
BT021124.1 | - | 1 - 2 | 328 | 90 | ||
BT025258.1 | - | 1 - 2 | 229 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |