Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119233.2 | NP_567859.1 | 4 - 5 | 792 | 64 | |
GenBank mRNA | AY050894.1 | - | 4 - 5 | 764 | 64 | |
AY096675.1 | - | 4 - 5 | 690 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |