Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119233.2 | NP_567859.1 | 6 - 7 | 1006 | 65 | |
GenBank mRNA | AY050894.1 | - | 6 - 7 | 978 | 65 | |
AY096675.1 | - | 6 - 7 | 904 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |