Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119262.6 | NP_194841.2 | 16 - 17 | 1446 | 134 | |
NM_202918.2 | NP_974647.1 | 16 - 17 | 1387 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |