Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119262.6 | NP_194841.2 | 7 - 8 | 809 | 77 | |
NM_202918.2 | NP_974647.1 | 7 - 8 | 750 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |