Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNGC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345080.1 | NP_001332114.1 | 8 - 9 | 2263 | 160 | |
NM_124805.3 | NP_200236.1 | 8 - 9 | 2224 | 160 | ||
GenBank mRNA | AK316810.1 | - | 8 - 9 | 2220 | 160 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |