Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRE11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345081.1 | NP_001330550.1 | 1 - 2 | 160 | 146 | |
NM_124806.2 | NP_200237.1 | 3 - 4 | 346 | 146 | ||
GenBank mRNA | AJ243822.1 | - | 3 - 4 | 315 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |