Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124811.5 | NP_568807.1 | 6 - 7 | 778 | 118 | |
GenBank mRNA | AK226402.1 | - | 6 - 7 | 586 | 118 | |
AY099716.1 | - | 6 - 7 | 724 | 118 | ||
BT000287.1 | - | 6 - 7 | 574 | 118 | ||
FJ794601.1 | - | 6 - 7 | 676 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |