Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124811.5 | NP_568807.1 | 9 - 10 | 1404 | 69 | |
GenBank mRNA | AK226402.1 | - | 9 - 10 | 1212 | 69 | |
AY099716.1 | - | 9 - 10 | 1350 | 69 | ||
BT000287.1 | - | 9 - 10 | 1200 | 69 | ||
FJ794601.1 | - | 9 - 10 | 1302 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |