Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGLU41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345089.1 | NP_001330205.1 | 6 - 7 | 846 | 63 | |
NM_001345090.1 | NP_001330204.1 | 9 - 10 | 1137 | 63 | ||
NM_124837.3 | NP_200268.3 | 9 - 10 | 1137 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |