Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345094.1 | NP_001330580.1 | 8 - 9 | 1257 | 95 | |
NM_001345095.1 | NP_001330581.1 | 6 - 7 | 881 | 95 | ||
NM_124848.3 | NP_568811.1 | 8 - 9 | 1376 | 95 | ||
GenBank mRNA | AK228138.1 | - | 8 - 9 | 1307 | 95 | |
BT003156.1 | - | 7 - 8 | 1028 | 95 | ||
D21138.1 | - | 8 - 9 | 1278 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |