Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124858.4 | NP_200288.1 | 2 - 3 | 1006 | 68 | |
GenBank mRNA | AF419604.1 | - | 2 - 3 | 930 | 68 | |
AF428355.1 | - | 2 - 3 | 930 | 68 | ||
AF428385.1 | - | 2 - 3 | 930 | 68 | ||
AY054216.1 | - | 2 - 3 | 930 | 68 | ||
AY058094.1 | - | 2 - 3 | 930 | 68 | ||
AY143082.1 | - | 2 - 3 | 891 | 68 | ||
U17589.1 | - | 2 - 3 | 912 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |