Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VTC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124894.5 | NP_200323.1 | 2 - 3 | 718 | 81 | |
GenBank mRNA | AY063788.1 | - | 2 - 3 | 526 | 81 | |
AY091285.1 | - | 2 - 3 | 210 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |