Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036999.1 | NP_001032076.1 | 3 - 4 | 378 | 96 | |
NM_124895.3 | NP_200324.1 | 3 - 4 | 511 | 96 | ||
GenBank mRNA | AF124159.1 | - | 3 - 4 | 428 | 96 | |
AK317295.1 | - | 3 - 4 | 375 | 96 | ||
BT029156.1 | - | 3 - 4 | 401 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |