Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036999.1 | NP_001032076.1 | 9 - 10 | 861 | 79 | |
NM_001345117.1 | NP_001330104.1 | 7 - 8 | 676 | 79 | ||
NM_124895.3 | NP_200324.1 | 9 - 10 | 994 | 79 | ||
GenBank mRNA | AF124159.1 | - | 9 - 10 | 911 | 79 | |
AK317295.1 | - | 9 - 10 | 858 | 79 | ||
BT029156.1 | - | 9 - 10 | 884 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |