Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOP1BETA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124913.4 | NP_200342.1 | 12 - 13 | 2466 | 88 | |
GenBank mRNA | AK226832.1 | - | 12 - 13 | 2382 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |