Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203616.2 | NP_001190545.1 | 13 - 14 | 2284 | 114 | |
NM_001345123.1 | NP_001332152.1 | 12 - 13 | 2552 | 114 | ||
NM_124921.5 | NP_200350.2 | 13 - 14 | 2339 | 114 | ||
GenBank mRNA | KF112069.1 | - | 12 - 13 | 2161 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |