Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GEX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345127.1 | NP_001318802.1 | 4 - 5 | 1274 | 122 | |
NM_001345129.1 | NP_001330007.1 | 4 - 5 | 1296 | 122 | ||
NM_001345132.1 | NP_001330006.1 | 3 - 4 | 1270 | 122 | ||
NM_001345133.1 | NP_001330005.1 | 4 - 5 | 1296 | 122 | ||
NM_001345134.1 | NP_001330002.1 | 4 - 5 | 1296 | 122 | ||
NM_124931.2 | NP_200360.2 | 3 - 4 | 1270 | 122 | ||
GenBank mRNA | AK175610.1 | - | 3 - 4 | 1270 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |