Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124941.3 | NP_200370.1 | 4 - 5 | 1256 | 67 | |
GenBank mRNA | DQ979876.1 | - | 4 - 5 | 999 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |