Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124975.4 | NP_200404.1 | 3 - 4 | 892 | 115 | |
GenBank mRNA | AY070725.1 | - | 3 - 4 | 781 | 115 | |
BT004528.1 | - | 3 - 4 | 619 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |