Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345176.1 | NP_001332655.1 | 6 - 7 | 900 | 79 | |
NM_124981.4 | NP_200410.1 | 6 - 7 | 900 | 79 | ||
GenBank mRNA | AF076252.1 | - | 5 - 6 | 438 | 79 | |
AY052304.1 | - | 5 - 6 | 483 | 79 | ||
AY139808.1 | - | 4 - 5 | 374 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |