Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAFII15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342109.1 | NP_001329548.1 | 6 - 7 | 717 | 99 | |
NM_119321.4 | NP_194900.1 | 6 - 7 | 608 | 99 | ||
NM_179150.4 | NP_849481.1 | 6 - 7 | 591 | 99 | ||
GenBank mRNA | AF344879.1 | - | 6 - 7 | 518 | 99 | |
AF352813.1 | - | 6 - 7 | 518 | 99 | ||
AK317292.1 | - | 6 - 7 | 523 | 99 | ||
AY086560.1 | - | 6 - 7 | 523 | 99 | ||
AY463628.1 | - | 5 - 6 | 331 | 99 | ||
Y12904.1 | - | 508 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |