Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036690.1 | NP_001031767.1 | 11 - 12 | 1437 | 64 | |
NM_001203956.1 | NP_001190885.1 | 11 - 12 | 1394 | 64 | ||
NM_119351.3 | NP_194927.1 | 10 - 11 | 1456 | 64 | ||
NM_179152.3 | NP_849483.1 | 11 - 12 | 1469 | 64 | ||
GenBank mRNA | AK316993.1 | - | 11 - 12 | 1322 | 64 | |
AK317084.1 | - | 10 - 11 | 1457 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |