Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179180.3 | NP_849511.1 | 6 - 7 | 1274 | 94 | |
GenBank mRNA | BT029750.1 | - | 6 - 7 | 1146 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |