Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204022.1 | NP_001190951.1 | 11 - 12 | 795 | 81 | |
NM_119986.3 | NP_195537.2 | 11 - 12 | 819 | 81 | ||
NM_179186.1 | NP_849517.1 | 11 - 12 | 819 | 81 | ||
GenBank mRNA | AJ243198.1 | - | 11 - 12 | 733 | 81 | |
AY099838.1 | - | 11 - 12 | 790 | 81 | ||
BT000334.1 | - | 11 - 12 | 703 | 81 | ||
KJ138715.1 | - | 11 - 12 | 703 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |